missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bis(4-nitrophenyl) Phosphate 98.0+%, TCI America™

Catalog No. B10991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B10991G Supplier TCI America Supplier No. B10991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4043-96-3
Molecular Formula C12H8N2NaO8P
Molecular Weight (g/mol) 362.166
MDL Number MFCD00065378
InChI Key DELHRHCJGSTQNU-UHFFFAOYSA-M
Synonym Phosphoric Acid Bis(4-nitrophenyl) Ester Sodium Salt
PubChem CID 3014253
IUPAC Name sodium;bis(4-nitrophenyl) phosphate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OP(=O)([O-])OC2=CC=C(C=C2)[N+](=O)[O-].[Na+]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 362.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Bis(4-nitrophenyl) Phosphate [for Phosphodiesterase Substrate]