missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bis(2-methoxyethoxy)aluminum Dihydride (70% in Toluene, ca. 3.6mol/L), TCI America™

Catalog No. S0467500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0467500G 500 g
S046725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0467500G Supplier TCI America Supplier No. S0467500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22722-98-1
Molecular Formula C6H16AlNaO4
MDL Number MFCD00011631
Synonym Sodium Dihydridobis(2-methoxyethoxy)aluminate

Specifications

Color Yellow
Quantity 500 g
UN Number 3399
Formula Weight 202.16
Physical Form Liquid
Chemical Name or Material Sodium Bis(2-methoxyethoxy)aluminum Dihydride (70% in Toluene, ca. 3.6mol/L)
EINECSNumber (2)-3011
TSCA Yes