missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™

Catalog No. AC430918000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430918000 800 mL AcroSeal™ Glass Bottle
AC430911000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430918000 Supplier Thermo Scientific Chemicals Supplier No. 430918000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22722-98-1 ∣ C6H16AlNaO4 ∣ 202.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 22722-98-1 , 108-88-3
Molecular Formula C6H16AlNaO4
Molecular Weight (g/mol) 202.16
MDL Number MFCD00011631
Synonym Sodium dihydro-bis-(2-methoxyethoxy)aluminate in toluene

Specifications

Boiling Point 110.0°C
Color Amber to Colorless
Physical Form Viscous Liquid
Quantity 800 mL
Flash Point 4°C
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.036
Density 1.0360g/mL
Linear Formula [(CH3OCH2CH2O)2AlH2]Na
Merck Index 15,8702
Solubility Information Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers
Formula Weight 202.16
Percent Purity 68.5 to 71.5%
Grade Ungraded
Chemical Name or Material Sodium bis(2-methoxyethoxy)Aluminum hydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May be fata
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 245-178-2

RUO – Research Use Only