missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate-Sodium Chloride Solution, LabChem™

Catalog No. lc229482
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC229482 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC229482 Supplier LabChem Supplier No. LC229482
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5 , 144-55-8
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Colorless
CAS Max % 99.33, 0.59, 0.08
Physical Form Liquid
Quantity 1 L
Linear Formula NaHCO3
Solubility Information Soluble in water
Formula Weight 84.01
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride
Chemical Name or Material Sodium Bicarbonate-Sodium Chloride Solution
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature