missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, Saturated, for Neutralization, Certified, LabChem™

Catalog No. LC229471
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC229471 500 mL
LC229474 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC229471 Supplier LabChem Supplier No. LC229471

Chemical Identifiers

CAS 144-55-8 , 7732-18-5
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Color Colorless
CAS Max % 90.4, 9.6
Linear Formula NaHCO3
Density 1.1g/mL
Solubility Information Soluble in water
Formula Weight 84.01
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.1g/mL
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Bicarbonate, Saturated, for Neutralization
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature