missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicarbonate de sodium (poudre/USP/FCC), Fisher Chemical™
Spécifications
Spécifications
| Nom ou substance chimique | Sodium Bicarbonate |
| Point de fusion | 270 °C |
| Couleur | Blanc |
| pH | 8.3 |
| Forme physique | Poudre/Solide |
| Quantité | 3 kg |
| Désignation | Test de réussite (FCC), Test de réussite (USP), Test de réussite (USP/FCC) |
| Perte à la dessication | 0,25% max. (FCC), 0,25% max. (USP), 0,25% max. (USP/FCC) |
| Emballage | Bouteille en polyéthylène |
| Renseignements sur la décomposition | >50 °C |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes and skin. May cause adverse kidney effects. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:2 Flammability:0 Instability:1 |