missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, 1M buffer soln., pH 8.5

Catalog No. AAJ60408AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60408AP 500 mL
AAJ60408AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60408AP Supplier Thermo Scientific Chemicals Supplier No. J60408AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong acids, strong oxidizing agents.

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 8.5
Physical Form Liquid
Quantity 500 mL
Solubility Information Soluble in water
Chemical Name or Material Sodium bicarbonate buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only