missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzohydroxamate Hydrate 98.0+%, TCI America™

Catalog No. B011225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B011225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B011225G Supplier TCI America Supplier No. B011225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 22513-32-2
Color White-Yellow
Molecular Formula C7H8NNaO3
Quantity 25 g
MDL Number MFCD00058967
Synonym Benzohydroxamic Acid Sodium Salt
InChI Key CTXNFHAZPJAZQN-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)N[O-].O.[Na+]
IUPAC Name sodium;N-oxidobenzamide;hydrate
Molecular Weight (g/mol) 177.135
PubChem CID 56924513
Formula Weight 159.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Benzohydroxamate Hydrate
Show More Show Less