missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide 99.0+%, TCI America™

Catalog No. S0489100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0489100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0489100G Supplier TCI America Supplier No. S0489100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
UN Number 1687
Formula Weight 65.01
Percent Purity ≥99.0% (T)
Chemical Name or Material Sodium Azide
EINECSNumber (1)-0482
RTECSNumber JY8050000
TSCA Yes