missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, For Chromium, 0.5% (w/v), LabChem™

Catalog No. LC229401
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC229401 500 mL
LC229402 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC229401 Supplier LabChem Supplier No. LC229401

Chemical Identifiers

CAS 26628-22-8 , 7732-18-5
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Color Colorless
CAS Max % 99.5, 0.5
Linear Formula NaN3
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 65.01
Percent Purity 0.5% (w/v)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Nitrogen oxides
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Azide, For Chromium
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature