missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Azide, 5% (w/v), Ricca Chemical
$247.16 - $247.16
Chemical Identifiers
| CAS | 26628-22-8 |
|---|---|
| Molecular Formula | N3Na |
| Molecular Weight (g/mol) | 65.011 |
| InChI Key | PXIPVTKHYLBLMZ-UHFFFAOYSA-N |
| Synonym | sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german |
| PubChem CID | 33557 |
| ChEBI | CHEBI:278547 |
| IUPAC Name | sodium;azide |
| SMILES | [N-]=[N+]=[N-].[Na+] |
Chemical Identifiers
| 26628-22-8 | |
| 65.011 | |
| sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
| CHEBI:278547 | |
| [N-]=[N+]=[N-].[Na+] |
| N3Na | |
| PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
| 33557 | |
| sodium;azide |
Specifications
| 26628-22-8 , 7732-18-5 | |
| 97.0,4.86 | |
| sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
| [N-]=[N+]=[N-].[Na+] | |
| 65.011 | |
| CHEBI:278547 | |
| Laboratory | |
| Alkaline | |
| Sodium Azide |
| 93.0,4.67 | |
| N3Na | |
| PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
| sodium;azide | |
| 33557 | |
| 5% | |
| Natural Poly Bottle | |
| 500 mL |