missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsénite de sodium, qualité réactif, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Sodium Arsenite |
| Couleur | Blanc |
| CAS % maximal | 100 |
| Forme physique | Poudre |
| Quantité | 100 g |
| Formule linéaire | NaAsO2 |
| Numéro UN | UN2027 |
| Renseignements sur la solubilité | Soluble in water |
| Poids de formule | 129.91 |
| Niveau | Réactif |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed or in contact with skin. Toxic if inhaled. May cause cancer (oral, inhalation). Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust. Do not get in eyes, on skin, or on clothing. Do not eat, drink or smoke when using this product. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Use only outdoors or in a well-ventilated area. Avoid release to the environment. Store locked up in a well-ventilated place. Keep container tightly closed. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Immediately call a poison center or doctor. If on skin: Wash with plenty of soap and water. Immediately call a poison center/doctor. Take off immediately all contaminated clothing and wash it before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor. Collect spillage. Dispose of contents/container to comply with local, state and federal regulations. |
| EINECSNumber | 232-070-5 |
| Recommended Storage | Température ambiante |