missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenite, For Residual Chlorine, Certified, 0.100N ±0.001N (0.05M), LabChem™

Catalog No. LC229002
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC229002 1 L
LC229001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC229002 Supplier LabChem Supplier No. LC229002

Chemical Identifiers

CAS 7784-46-5 , 7732-18-5
Molecular Formula AsNaO2
Molecular Weight (g/mol) 129.909
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
Synonym sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
PubChem CID 443495
ChEBI CHEBI:29678
IUPAC Name sodium;oxoarsinite
SMILES [O-][As]=O.[Na+]

Specifications

Color Colorless
CAS Max % 99.35, 0.65
Linear Formula NaAsO2
Concentration 0.100N ±0.001N (0.05M)
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 129.91
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Arsenic and its oxides
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium Arsenite, For Residual Chlorine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause cancer (oral, inhalation).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, protective clothing, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature