missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenite, For Residual Chlorine, Certified, 0.100N ±0.001N (0.05M), LabChem™

Catalog No. LC229001
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC229001 500 mL
LC229002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC229001 Supplier LabChem Supplier No. LC229001

Chemical Identifiers

CAS 7784-46-5 , 7732-18-5
Molecular Formula AsNaO2
Molecular Weight (g/mol) 129.909
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
Synonym sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
PubChem CID 443495
ChEBI CHEBI:29678
IUPAC Name sodium;oxoarsinite
SMILES [O-][As]=O.[Na+]

Specifications

Color Colorless
CAS Max % 99.35, 0.65
Linear Formula NaAsO2
Concentration 0.100N ±0.001N (0.05M)
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 129.91
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Arsenic and its oxides
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Arsenite, For Residual Chlorine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause cancer (oral, inhalation).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, protective clothing, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature