missing translation for 'onlineSavingsMsg'
Learn More

Sodium aluminum oxide, tech.

Catalog No. AA3545336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3545336 500 g
AA3545322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3545336 Supplier Thermo Scientific Chemicals Supplier No. 03545336
Only null left

CAS: 1302-42-7 | Al2O3·Na2O

Sodium aluminum oxide is used in the isomerization of alkene and isomerization of amines. It is also used in water treatment as an adjunct and as a coagulant aid to flocculation. It is involved in the removal of silica and phosphates from waste water. It is employed to accelerate concrete solidification. It finds application in the paper industry and fire brick production. It acts as an intermediate in the production of zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1302-42-7
Molecular Formula Al2O3·Na2O
MDL Number MFCD00801010
Synonym Sodium aluminate

Specifications

Color White
Physical Form Crystalline
Quantity 500 g
Solubility Information Insoluble in alcohol,soluble in waterInsoluble in alcohol. Soluble in water.
Formula Weight 163.94
Sensitivity Hygroscopic
Appearance White crystalline
Density 1.52
Chemical Name or Material Sodium aluminum oxide
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packing Group: III
EINECSNumber 215-100-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only