missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, ACS Grade, 99.0% to 101.0%, LabChem™

Catalog No. LC228201
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC228201 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC228201 Supplier LabChem Supplier No. LC228201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Boiling Point 123°C
Melting Point 58°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula CH3COOHNa·3H2O
Solubility Information Soluble in water
Formula Weight 136.08
Percent Purity 99.0 to 101.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.45g/cm3
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Density 1.45g/cm3
Chemical Name or Material Sodium Acetate, Trihydrate
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature