missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, For Zinc, Certified, 2.0M ±0.1M (2.0N), LabChem™

Catalog No. LC228504
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC228504 4 L
LC228501 500 mL
LC228502 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC228504 Supplier LabChem Supplier No. LC228504

Chemical Identifiers

CAS 6131-90-4 , 7732-18-5
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Color Colorless
CAS Max % 76, 24
Linear Formula CH3COOHNa·3H2O
Concentration 2.0M ±0.1M (2.0N)
Density 1.13g/mL
Solubility Information Soluble in water
Formula Weight 136.08
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.13g/mL
Traceability to NIST Traceable to NIST
Viscosity 3.017cSt
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Sodium Acetate, For Zinc
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature