Learn More
Sodium acetate, anhydrous, 99.997% (metals basis)
Description
Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 324°C |
| pH | 7.5 to 9 |
| Physical Form | Powder |
| Quantity | 100 g |
| Sensitivity | Hygroscopic |
| Density | 1.528 |
| Beilstein | 3595639 |
| Merck Index | 14,8571 |
| Solubility Information | Very soluble in water. Soluble in alcohol. |
| Formula Weight | 82.03 |
| Show More |
Safety and Handling
| Hazard Category | H500 |
| EINECSNumber | 204-823-8 |
| RTECSNumber | AJ4300010 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only