missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium AA Standard, 1000 ppm Na in H2O, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS Min % | 98.0, 0.25 |
| CAS Max % | 100.0, 0.26 |
| Odor | Odorless |
| Color | Colorless |
| Boiling Point | 100°C |
| Type | Standard |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | 1mL = 1mg Na (1, 000ppm Na)NaCl in Water |
| Chemical Name or Material | Sodium AA Standard |
Safety and Handling
| ShelfLife | 24 months |