missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 8-Ethoxyquinoline-5-sulfonate Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 275.25 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium 8-Ethoxyquinoline-5-sulfonate Hydrate |
Safety and Handling
| TSCA | No |