missing translation for 'onlineSavingsMsg'
Learn More

Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate 98.0+%, TCI America™

Catalog No. S09765G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09765G 5 g
S09761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09765G Supplier TCI America Supplier No. S09765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6223-35-4
Molecular Formula C15H17NaO3S
Molecular Weight (g/mol) 300.348
MDL Number MFCD00866167
InChI Key GEYJUFBPCGDENK-UHFFFAOYSA-M
Synonym 7-Isopropyl-1,4-dimethyl-azulene-3-sulfonic Acid Sodium Salt, Sodium Gualenate, Sodium 5-Isopropyl-3,8-dimethylazulene-1-sulfonate, 5-Isopropyl-3,8-dimethylazulene-1-sulfonic Acid Sodium Salt, Sodium Guaiazulenesulfonate, Guaiazulenesulfonic Acid Sodium S
PubChem CID 23669592
IUPAC Name sodium;3,8-dimethyl-5-propan-2-ylazulene-1-sulfonate
SMILES CC1=C2C(=CC(=C2C=C(C=C1)C(C)C)C)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 98°C
Color Blue
Quantity 5 g
Formula Weight 300.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 7-Isopropyl-1,4-dimethylazulene-3-sulfonate
EINECSNumber (9)-0414
RTECSNumber CO4826000
TSCA No