missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Aminophenylarsonate 98.0+%, TCI America™

Catalog No. A053125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A053125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A053125G Supplier TCI America Supplier No. A053125G
Only null left

Chemical Identifiers

CAS 127-85-5
Molecular Formula C6H7AsNNaO3
Molecular Weight (g/mol) 239.04
MDL Number MFCD09750377
InChI Key OUFRIWNNMFWZTM-UHFFFAOYSA-M
Synonym 4-Aminobenzenearsonic Acid Sodium Salt, p-Arsanilic Acid Sodium Salt, Atoxyl, 4-Aminophenylarsonic Acid Sodium Salt, Sodium Arsanilate, Sodium Hydrogen 4-Aminophenylarsonate
PubChem CID 131864343
IUPAC Name sodium hydrogen (4-aminophenyl)arsonate
SMILES [Na+].NC1=CC=C(C=C1)[As](O)([O-])=O

Specifications

Quantity 25 g
UN Number 2473
Formula Weight 239.04
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Aminophenylarsonate
RTECSNumber CF9625000
TSCA No