missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-(N-Ethylanilino)propanesulfonate 98.0+%, TCI America™

Catalog No. S0817200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0817200MG 200 mg
S08171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0817200MG Supplier TCI America Supplier No. S0817200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82611-85-6
Molecular Formula C11H16NNaO3S
Molecular Weight (g/mol) 265.303
MDL Number MFCD01311047
InChI Key FFJBIXKLISICDT-UHFFFAOYSA-M
Synonym 3-(N-Ethylanilino)propanesulfonic Acid Sodium Salt, N-Ethyl-N-(3-sulfopropyl)aniline Sodium Salt, ALPS
PubChem CID 23685294
IUPAC Name sodium;3-(N-ethylanilino)propane-1-sulfonate
SMILES CCN(CCCS(=O)(=O)[O-])C1=CC=CC=C1.[Na+]

Specifications

Color White
Quantity 200 mg
Formula Weight 265.30
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-(N-Ethylanilino)propanesulfonate [for Biochemical Research]