missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3,5-Dichloro-2-hydroxybenzenesulfonate 99.0+%, TCI America™

Catalog No. D192825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D192825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D192825G Supplier TCI America Supplier No. D192825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54970-72-8
Molecular Formula C6H3Cl2NaO4S
Molecular Weight (g/mol) 265.036
MDL Number MFCD00009798
InChI Key NMWCVZCSJHJYFW-UHFFFAOYSA-M
Synonym sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
PubChem CID 3085077
IUPAC Name sodium;3,5-dichloro-2-hydroxybenzenesulfonate
SMILES C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]

Specifications

Quantity 25 g
Formula Weight 265.04
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3,5-Dichloro-2-hydroxybenzenesulfonate [for Biochemical Research]