missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Oxobutyrate 98.0+%, TCI America™

Catalog No. K00405G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K00405G Supplier TCI America Supplier No. K00405G
Only null left

Chemical Identifiers

CAS 2013-26-5
Molecular Formula C4H5NaO3
Molecular Weight (g/mol) 124.07
MDL Number MFCD00064194
InChI Key SUAMAHKUSIHRMR-UHFFFAOYSA-M
Synonym 2-Oxobutyric Acid Sodium Salt, Sodium 2-Ketobutyrate, 2-Ketobutyric Acid Sodium Salt
PubChem CID 23673454
IUPAC Name sodium 2-oxobutanoate
SMILES [Na+].CCC(=O)C([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 124.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Oxobutyrate
RTECSNumber ET5955900
TSCA Yes