missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Mercaptoethanesulfonate 95.0+%, TCI America™

Numéro de catalogue. M091310G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M091310G 10 g
M091325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M091310G Fournisseur TCI America Code fournisseur M091310G
Il en reste null

Identifiants chimiques

CAS 19767-45-4
Molecular Formula C2H5NaO3S2
Molecular Weight (g/mol) 164.169
MDL Number MFCD00007535
InChI Key XOGTZOOQQBDUSI-UHFFFAOYSA-M
Synonym mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
PubChem CID 23662354
IUPAC Name sodium;2-sulfanylethanesulfonate
SMILES C(CS(=O)(=O)[O-])S.[Na+]

Spécifications

Color White
Quantity 10 g
Formula Weight 164.17
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Mercaptoethanesulfonate
RTECSNumber KI7968000
TSCA No