missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,4-Dichlorophenoxyacetate Monohydrate 98.0+%, TCI America™

Catalog No. D131925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D131925G 25 g
D1319500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D131925G Supplier TCI America Supplier No. D131925G

Chemical Identifiers

CAS 2702-72-9
Molecular Formula C8H8Cl2NaO4
Molecular Weight (g/mol) 262.038
MDL Number MFCD00068284
InChI Key JXHUJQWKJUFZSW-UHFFFAOYSA-N
Synonym 2,4-Dichlorophenoxyacetic Acid Sodium Salt, Na-2,4-D
PubChem CID 124204223
IUPAC Name 2-(2,4-dichlorophenoxy)acetic acid;sodium;hydrate
SMILES C1=CC(=C(C=C1Cl)Cl)OCC(=O)O.O.[Na]

Specifications

Melting Point 216°C
Quantity 25 g
Formula Weight 243.01
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,4-Dichlorophenoxyacetate Monohydrate
EINECSNumber (3)-0927
RTECSNumber AG8925000
TSCA Yes