missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2,3,5,6-Tetrafluoro-4-hydroxybenzenesulfonate 98.0+%, TCI America™

Catalog No. S08365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08365G 5 g
S08361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08365G Supplier TCI America Supplier No. S08365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 221908-17-4
Molecular Formula C6HF4NaO4S
Molecular Weight (g/mol) 268.11
MDL Number MFCD03427812
InChI Key HVXPVJLSYQBYCR-UHFFFAOYSA-M
Synonym 2,3,5,6-Tetrafluoro-4-hydroxybenzenesulfonic Acid Sodium Salt
PubChem CID 23686972
IUPAC Name sodium 2,3,5,6-tetrafluoro-4-hydroxybenzene-1-sulfonate
SMILES [Na+].OC1=C(F)C(F)=C(C(F)=C1F)S([O-])(=O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 268.11
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2,3,5,6-Tetrafluoro-4-hydroxybenzenesulfonate