missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Tridecanesulfonate 98.0+%, TCI America™

Catalog No. I03515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03515G 5 g
I035125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03515G Supplier TCI America Supplier No. I03515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5802-89-1
Molecular Formula C13H27NaO3S
Molecular Weight (g/mol) 286.406
MDL Number MFCD00024993
InChI Key CACJZDMMUHMEBN-UHFFFAOYSA-M
Synonym IPC-ALKS-13, 1-Tridecanesulfonic Acid Sodium Salt
PubChem CID 23683718
IUPAC Name sodium;tridecane-1-sulfonate
SMILES CCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 286.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Tridecanesulfonate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-1640
TSCA No