missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-propanethiolate, tech. 85%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Gray to White |
| Odor | Stench |
| Linear Formula | CH3CH2CH2SNa |
| Quantity | 5 g |
| UN Number | UN3263 |
| Beilstein | 3556024 |
| Sensitivity | Air and moisture sensitive |
| Formula Weight | 98.14 |
| Percent Purity | 85% |
| Physical Form | Solid |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S. |
| EINECSNumber | 230-005-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only