missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Octanesulfonate 98.0+%, TCI America™

Catalog No. I03465G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03465G 5 g
I034625G 25 g
I0346100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I03465G Supplier TCI America Supplier No. I03465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium 1-Octanesulfonate [Reagent for Ion-Pair Chromatography], >-98.0% (T), C8H17NaO3S, CAS Number-5324-84-5

Sodium 1-Octanesulfonate [Reagent for Ion-Pair Chromatography], >-98.0% (T), C8H17NaO3S, CAS Number-5324-84-5

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 216.27
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium 1-Octanesulfonate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-1640
TSCA Yes