missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Naphthaleneacetate 96.0+%, TCI America™

Catalog No. N0007500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0007500G 500 g
N000725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0007500G Supplier TCI America Supplier No. N0007500G

Chemical Identifiers

CAS 61-31-4
Molecular Formula C12H9NaO2
Molecular Weight (g/mol) 208.192
MDL Number MFCD00064967
InChI Key CJUUXVFWKYRHAR-UHFFFAOYSA-M
Synonym 1-Naphthylacetic Acid Sodium Salt, 1-Naphthaleneacetic Acid Sodium Salt, Sodium 1-Naphthylacetate, Na-NAA
PubChem CID 23694671
ChEBI CHEBI:81811
IUPAC Name sodium;2-naphthalen-1-ylacetate
SMILES C1=CC=C2C(=C1)C=CC=C2CC(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 208.19
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Naphthaleneacetate
EINECSNumber (4)-0410
RTECSNumber QJ1355000
TSCA Yes