missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Dodecanesulfonate 98.0+%, TCI America™

Catalog No. D12225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12225G 5 g
D122225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12225G Supplier TCI America Supplier No. D12225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2386-53-0
Molecular Formula C12H25NaO3S
Molecular Weight (g/mol) 272.379
MDL Number MFCD00007527
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
PubChem CID 23665726
IUPAC Name sodium;dodecane-1-sulfonate
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 272.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Dodecanesulfonate
EINECSNumber (2)-1640
TSCA Yes