missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl SmartCell™ Post Mount Divider
Description
- Allow for custom compartmentalization of the SmartCell™ basket system.
- Large and small articles alike are stored in separate compartments, allowing for easy identification and quick retrieval.
Spécifications
Spécifications
| Color | Chrome |
| Material | Steel |
| For Use With (Application) | Shelving |
| Shelf Type | Divider |
| Width (English) | 10 in. |