missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl SmartCell™ Clear Wire Shelf Label Holder
Description
- Add-on option for additional customization
- Clear
- Three ways to mount
Spécifications
Spécifications
| Color | Chrome |
| Material | Steel |
| For Use With (Application) | Shelving |
| For Use With (Equipment) | SmartCell™ |
| Shelf Type | Basket Label Holder |
| Width (English) | 36 in. |