Learn More
Thermo Scientific™ Smart-Vue Pro Cloud Software and Subscription Services
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
Supplier:  Thermo Scientific™ SVPSWCS21000
 Afficher plus de versions de ce produit
Numéro de catalogue. SVSWCS21000
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description
Description
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times.
  • Intuitive dashboard simplifies daily usage.
  • Optional floor plan and sites views.
  • Data can be exported with a click for reporting and customizable graphs.
  • Web application on Cloud or customer server.
  • Instant access to alarms, readings, and graphs.
  • Remote data access.
  • Flexible organization by site, department user, equipment, and criticality.
  • And trail and secure-level access control to support 21CFR Part 11
  • Cloud-based configurations allow flexibility to check status anytime, anywhere from any modern web browser.
Spécifications
Identifiants chimiques
Smart-Vue Pro Software and Subscription Service
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
Spécifications
Smart-Vue Pro Software and Subscription Service
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
Smart-Vue Pro Cloud license key - Access to the Cloud and Smart-Vue Pro Web application for installation up to 1000 sensors. License validity: 12 months after activation
Up to 1000 sensors
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS