Learn More
Thermo Scientific™ Smart-Vue Pro Cloud Software and Subscription Services
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
Supplier: Thermo Scientific™ SVPSWCS20020
Description
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times.
- Intuitive dashboard simplifies daily usage.
- Optional floor plan and sites views.
- Data can be exported with a click for reporting and customizable graphs.
- Web application on Cloud or customer server.
- Instant access to alarms, readings, and graphs.
- Remote data access.
- Flexible organization by site, department user, equipment, and criticality.
- And trail and secure-level access control to support 21CFR Part 11
- Cloud-based configurations allow flexibility to check status anytime, anywhere from any modern web browser.
Identifiants chimiques
| Smart-Vue Pro Software and Subscription Service | |
| Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module |
Spécifications
| Smart-Vue Pro Software and Subscription Service | |
| Smart-Vue Pro Cloud license key - Access to the Cloud and Smart-Vue Pro Web application for installation up to 20 sensors. License validity: 12 months after activation | |
| Up to 20 sensors | |
| Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module |