Learn More
Thermo Scientific™ Smart-Vue Pro Accessories
Catalog No. SVACCBAT002
Change view
Click to view available options
Type:
Adapter
Battery
For Use With (Equipment):
2 or 3 Wire Generic PT100 Probes
Smart-Vue Classic PT100 Probes
Smart-Vue Pro Duo or Quatro Module
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
SVACCBAT002 Battery Smart-Vue Pro Duo or Quatro Module
11676460 Adapter 2 or 3 Wire Generic PT100 Probes
11676461 Adapter Smart-Vue Classic PT100 Probes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SVACCBAT002 Supplier Thermo Scientific™ Supplier No. SVPACCBAT002
Customize your Smart-Vue Pro remote monitoring system with accessories to meet your specific needs.
Choose from an assorment of Smart-Vue Pro accessories including power supplies, cables and mounting hardware, antenna and audible alarms to customize your remote monitoring system as well as ensure system integrity.
Compatible with Smart-Vue Pro remote monitoring system.
Specifications
Type Battery
Battery Type 2 Batteries 3.6 V Batteries
Description Smart-Vue Pro 3.6 V Lithium Module Battery (Pk. of 2)
Includes 3.6 V Lithium Batteries - Qty. 2
Certifications/Compliance Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
For Use With (Application) Smart-Vue Pro Remote Monitoring System
For Use With (Equipment) Smart-Vue Pro Duo or Quatro Module