missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMARCC1 Recombinant Rabbit Monoclonal Antibody (23GB4165)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553700
Description
SMARCC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and contains a predicted leucine zipper motif typical of many transcription factors.Chemical Identifiers
| SMARCC1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| SMARCC1 | |
| AI115498; BAF155; BRG1-associated factor 155; chromatin remodeling complex BAF155 subunit; CRACC1; mammalian chromatin remodeling complex BRG1-associated factor 155; Rsc8; Smarcc1; Srg3; SWI/SNF complex 155 kDa subunit; SWI/SNF complex subunit SMARCC1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1; SWI3; SWI3-related protein | |
| SMARCC1 | |
| Primary | |
| Affinity chromatography |
| 23GB4165 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97496, Q92922 | |
| 20588, 301020, 6599 | |
| A synthesized peptide derived from SMARCC1/BAF155 (700-750AA). | |
| Antibody |
Specifications
| SMARCC1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SMARCC1 | |
| AI115498; BAF155; BRG1-associated factor 155; chromatin remodeling complex BAF155 subunit; CRACC1; mammalian chromatin remodeling complex BRG1-associated factor 155; Rsc8; Smarcc1; Srg3; SWI/SNF complex 155 kDa subunit; SWI/SNF complex subunit SMARCC1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1; SWI3; SWI3-related protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20588, 301020, 6599 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4165 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97496, Q92922 | |
| SMARCC1 | |
| A synthesized peptide derived from SMARCC1/BAF155 (700-750AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |