missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Smallpox Virus B5R Monoclonal Antibody (1J2F3)
GREENER_CHOICE

Numéro de catalogue. 01676594
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
01676594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01676594 Fournisseur Invitrogen™ Code fournisseur MA141212

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

This antibody is predicted to react with Bovinepox virus, Ectromelia virus, Monkeypox virus, Rabbitpox virus, and vaccinia virus. Immunogen sequence: IQEYNLIRTY KKVDK.

B5R is an EEV-specific Type I membrane glycoprotein essential for EEV formation. Vaccinia virus, a member of Poxviridae family, is a dsDNA enveloped virus. It consists of 4 types of virion stages among which extracellular enveloped virus (EEV) is critical for cell-to-cell and long range viral infection. Some cellular antigens are associated with EEV and these vary with the cell type used to grow the virus. B5R consists of a large ectodomain containing regions similar to SCR domains of complement regulatory proteins followed by a transmembrane region and a cytoplasmic tail. B5R is essential for efficient wrapping of IMV, actin tail formation, normal plaque size, virus virulence, and for EEV formation.

Spécifications

Antigène Smallpox Virus B5R
Applications Western Blot
Classification Monoclonal
Clone 1J2F3
Concentration 1.0 mg/mL
Conjugué Unconjugated
Formule PBS with 0.05% sodium azide
Espèces hôtes Mouse
Immunogène Amino acids 33-47 of the B5R protein.
Méthode de purification Protein G
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Virus
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus de résultats Afficher moins de résultats