missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Small Volume™ 384 Well Polystyrene Flat Bottom Microplates

Provides ultra low non-specific biomolecular binding properties.

$1,556.14 - $1,698.29

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Autoclavable Not Autoclavable
Material Polystyrene
View More Specs

Products 2
Catalog Number Mfr. No. Color Surface Treatment Price Quantity  
Catalog Number Mfr. No. Color Surface Treatment Price Quantity  
07-000-032
View Documents
Greiner Bio-One
784101
Clear Non-treated N/A
Only null left
 
07000077
View Documents
Greiner Bio-One
784904
White Protein-Repellent Non-binding N/A
Only null left
 
Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Long-term surface stability without degradation or leaching
  • Higher assay sensitivity with reduced background
Specifications

Specifications

Standard
16 x 24
Polystyrene
4 to 25 μL
28 μL
800 x g (swinging-bucket rotor)
10
-20°C to +60°C
Without Lid
Not Autoclavable
384
Non-sterile
Yes
40
Yes
Clear
Videos
SDS
Documents

Documents