missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Small Volume™ 384 Well Polystyrene Flat Bottom Microplates
Provides ultra low non-specific biomolecular binding properties.
Supplier: Greiner Bio-One 784904
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
Specifications
| Standard | |
| 16 x 24 | |
| White | |
| 384 | |
| Non-sterile | |
| 28 μL | |
| 800 x g (swinging-bucket rotor) | |
| 10 | |
| -20°C to +60°C |
| Without Lid | |
| Not Autoclavable | |
| Polystyrene | |
| 4 to 25 μL | |
| Protein-Repellent Non-binding | |
| Yes | |
| 40 | |
| Yes | |
| Clear |