missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Small Volume™ 384 Well Polystyrene Flat Bottom Microplates

Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 4 to 25 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Show More |