Learn More
Invitrogen™ SMAD3 Monoclonal Antibody (E.980.9)
GREENER_CHOICE
Rabbit Monoclonal Antibody
Supplier:  Invitrogen™ MA514939
Numéro de catalogue. PIMA514939
Il en reste null
Description
Description
It is not recommended to aliquot this antibody.
SMAD3 belongs to the SMAD family of proteins. They are similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.
Spécifications
Identifiants chimiques
SMAD3
95 μg/mL
ChIP Assay, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P84022, P84025, Q8BUN5
17127, 25631, 4088
Synthetic peptide corresponding to residues at the amino terminus of Smad3.
Antibody
E.980.9
-20°C
Monoclonal
Liquid
IgG
Human, Mouse, Rat, Monkey
Smad3
AU022421; DKFZp586N0721; DKFZp686J10186; hMAD-3; hSMAD3; HSPC193; HsT17436; JV15-2; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 3; MAD homolog 3a; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MAD-3; madh3; madh3a; madh3-A; MGC60396; mMad3; mothers against decapentaplegic homolog 3; mothers against decapentaplegic homolog 3a; mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD; Smad 3; SMAD family member 3; SMAD family member 3 L homeolog; SMAD family member 3a; SMAD, mothers against DPP homolog 3; Smad3; smad3.L; smad3a; TGF beta response effector Smad3; TGF-beta response effector Smad3; transcription factor; transcription factor Smad3; wu:fa99e03; XELAEV_18018454mg; XenMLP; Xmad3; XSmad3
Smad3
Primary
Affinity chromatography
Spécifications
SMAD3
Monoclonal
95 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P84022, P84025, Q8BUN5
Smad3
Synthetic peptide corresponding to residues at the amino terminus of Smad3.
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG
ChIP Assay, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
E.980.9
Unconjugated
Smad3
AU022421; DKFZp586N0721; DKFZp686J10186; hMAD-3; hSMAD3; HSPC193; HsT17436; JV15-2; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 3; MAD homolog 3a; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MAD-3; madh3; madh3a; madh3-A; MGC60396; mMad3; mothers against decapentaplegic homolog 3; mothers against decapentaplegic homolog 3a; mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD; Smad 3; SMAD family member 3; SMAD family member 3 L homeolog; SMAD family member 3a; SMAD, mothers against DPP homolog 3; Smad3; smad3.L; smad3a; TGF beta response effector Smad3; TGF-beta response effector Smad3; transcription factor; transcription factor Smad3; wu:fa99e03; XELAEV_18018454mg; XenMLP; Xmad3; XSmad3
Rabbit
Affinity chromatography
RUO
17127, 25631, 4088
-20°C
Liquid
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS