missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Slipper Animalcule Paramecium Model

Catalog No. S27899
Change view
Click to view available options
Dimensions (L x W x H):
15.5 x 5.25 x 3.75 in. (Model)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S27899 15.5 x 5.25 x 3.75 in. (Model)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27899 Supplier Eisco™ Supplier No. EISZM0016

A 3D, sectional view of a Slipper Animalcule Paramecium with hand painted details and removable parts.

  • Greatly enlarged model of a Slipper Animalcule Paramecium
  • Key features include Pellicle, ectoplasm, endoplasm, trichocytes, peristome, vestibule, buccal cavity, gullet, cyto pharynx, food vacuole, macronucleus, micronucleus, contractile vacuole (relaxed, and expanded), opening of contractile vacuole, and cilia
  • Made of durable polyvinyl chloride (PVC) plastic which is corrosion resistant, lightweight, and has high strength
  • Resists reactions with acids, gasoline, alcohol, and hydrocarbons
  • Vividly hand painted with eco friendly paint
  • Identification key card included

Specifications

Dimensions (L x W x H) 15.5 x 5.25 x 3.75 in. (Model)
Dimensions (L x W) 18 x 8 in. (Base)
For Use With (Application) Biological Studies
Height (English) 4.75 in.
Includes Identification Key Card
Material PVC
Mounting Base
Product Type Model