Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for four inner-door, ultra-low temperature freezers.
Supplier:  Thermo Scientific™ 1950521
 View more versions of this product
Catalog No. 1950521
Get Thermo Scientific Accessories with Purchase
View all promotions
Only null left
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 4 shelf Series Freezers as well as well as previous generation Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
Warranty
Warranty varies by country, please contact us for details
Specifications
Chemical Identifiers
Silver
Specifications
3 in. Box Racks - Adjustable side access rack
Boxes (3 in.)
15 Boxes/Rack
5.3 in.
26.8 in.
Side Access Freezer Rack (Adjustable)
29.5 cm
Silver
Stainless Steel
TDE and TSX Series (4 shelves)
11.6 in.
68.1 cm
3 Tiers
13.5 cm
Product Suggestions
Product Suggestions