Learn More
Electron Microscopy Sciences Easy Dip™ Slide Staining Jar, White
Numéro de catalogue. 501899558
Encompass
EasyDip Slide Staining system jars resist attacks from most staining reagents but not phenols. Wide base for stability. 2" x 3" x 3"
Color: White
Numéro de catalogue. 501899558 Fournisseur Electron Microscopy Sciences Code fournisseur 71385W
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
EasyDip Slide Staining system jars resist attacks from most staining reagents but not phenols. Wide base for stability. 2" x 3" x 3"
Color: White