Learn More
United Scientific™ Slide Mailer With Cap, PP, Neutral
Description
- A convenient and safe solution for holding and transferring microscope slides. The inside walls feature slotted channels to keep slides separated and to make removal easier. The lids close securely to keep the slides safe.
- Accommodates microscope slides in two sizes:
- 75.0 mm x 25.0 mm x (0.8 to 1.2 mm)
- 76.0 mm x 26.0 mm x (0.8 to 1.2 mm)
- Securely hold slides: 3-place option with screw cap, and 5-place option with hinge cap
- Durable Polypropylene
- Holds two common slide sizes
Spécifications
Spécifications
| Type | Slide Mailer |
| Slide Capacity | 3 |
| Material | Polypropylene |