missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC39A9 Polyclonal Antibody

Description
A suggested positive control is HepG2 cell lysate. PA5-21074 can be used with blocking peptide PEP-1188.
The zinc transporter ZIP9, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP9 is a multipass membrane protein that belongs to the ZIP transporter subfamily I. It is located at the trans-Golgi network regardless of zinc presence and is thought to be a zinc homeostasis regulator acting in the secretory pathway without significantly altering cytosolic zinc homeostasis.
Spécifications
Spécifications
| Antigène | SLC39A9 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.02% sodium azide |
| Gène | SLC39A9 |
| Numéro d’acquisition du gène | Q8BFU1, Q9NUM3 |
| Alias du gène | 2010002A02Rik; 2610511I23Rik; 4833420E20Rik; AW209151; SLC39A9; solute carrier family 39 (zinc transporter), member 9; solute carrier family 39 member 9; solute carrier family 39, member 9; UNQ714/PRO1377; zinc transporter SLC39A9; zinc transporter ZIP9; ZIP9; ZIP-9; Zrt- and Irt-like protein 9 |
| Symboles de gène | SLC39A9 |
| Afficher plus de résultats |