missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC24A1 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA5101898
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA5101898 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA5101898 Fournisseur Invitrogen™ Code fournisseur PA5101898
Il en reste null

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC24A1.

SLC24A1 belongs to a family of potassium-dependent sodium/calcium exchangers. Members of this family have 2 large hydrophilic loops and 2 sets of multiple transmembrane-spanning segments (Schnetkamp, 2004 [PubMed 14770312]).

Spécifications

Antigen SLC24A1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC24A1
Gene Accession No. O60721
Gene Alias CSNB1D; HsT17412; KIAA0702; Na(+)/K(+)/Ca(2+)-exchange protein 1; NCKX; Nckx1; retinal Na/Ca,K exchanger; retinal rod Na+/Ca+/K+ exchanger; retinal rod Na-Ca+K exchanger; RODX; SLC24A1; sodium/calcium/potassium exchanger; sodium/potassium/calcium exchanger 1; solute carrier family 24 (sodium/potassium/calcium exchanger), member 1; solute carrier family 24 member 1
Gene Symbols SLC24A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC24A1(Accession O60721), corresponding to amino acid residues Y153-S203.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214111, 9187
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats